Cole-Parmer® Wet/Wet Differential Pressure Transmitters
These differential pressure transmitters are ideal for filter pressure drop monitoring and pumphead pressure applications
Capacitance sensor allows ports to be wet/wet, wet/dry, or dry/dry. Unique isolation system responds quickly to pressure changes—20 times faster than fluid-filled transmitters.
The rugged NEMA 4 (IP65) housing withstands harsh environments normally found in process control, liquid-level measurement, and energy management. All transmitters have screw terminal connections.
184,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Pressure (PSID) Wetted Materials
Availability Pricing
COMPARE
±0.25% full-scale 1 17-4PH SS, Viton®
₹184,000.00INR / Each
COMPARE
±0.25% full-scale 2.5 17-4PH SS, Viton®
₹184,000.00INR / Each
COMPARE
±0.25% full-scale 5 17-4PH SS, Viton®
₹184,000.00INR / Each
COMPARE
±0.25% full-scale 25 17-4PH SS, Viton®
₹184,000.00INR / Each