Cole-Parmer® Vertical/Chest Racks for Standard 3" Boxes
Use with a wide variety of chest freezers and liquid nitrogen tanks
  • Included locking rod keeps sample storage boxes secure within the rack
  • Dual metal handles for easy handling
  • Accommodates standard 3" cardboard, plastic, and metal boxes
  • Sturdy corrosion-resistant stainless steel construction
Racks feature sturdy handles for easy access to your samples in vertical/chest freezers.
7,700.00 - ₹27,400.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
4 boxes 12 3/4
₹7,700.00INR / Each
COMPARE
5 boxes 15 7/8
₹8,600.00INR / Each
COMPARE
6 boxes 19 1/8
₹10,800.00INR / Each
COMPARE
7 boxes 22 1/4
₹21,000.00INR / Each
COMPARE
8 boxes 25 7/16
₹22,500.00INR / Each
COMPARE
9 boxes 28 1/4
₹24,000.00INR / Each
COMPARE
10 boxes 31 3/4
₹27,400.00INR / Each
Showing 1- 7 of 7