Cole-Parmer® Vertical/Chest Half Racks for Standard 2" Boxes
Slim, lightweight design offers easy handling
  • Use with a variety of chest freezers and liquid nitrogen tanks
  • Accommodates standard 2" cardboard, plastic, and metal boxes
  • Sturdy corrosion-resistant stainless steel construction
  • Locking rod securely keeps boxes in place
Racks feature sturdy handles for easy access to your samples in vertical/chest freezers.
4,600.00 - ₹6,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
4 boxes 22
₹4,600.00INR / Each
COMPARE
5 boxes 26 1/2
₹6,300.00INR / Each