Cole-Parmer® Vertical/Chest Freezer Racks for 100-Cell Hinged Top Plastic Boxes
Conveniently store multiple 100-cell hinged top plastic boxes
  • Included locking rod securely holds boxes in place
  • Easily place and retrieve racks using sturdy dual handles
  • Corrosion-resistant stainless steel construction
Racks feature sturdy handles for easy access to your samples in vertical/chest freezers.
16,900.00 - ₹26,900.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
8 boxes 18 7/16
₹16,900.00INR / Each
COMPARE
10 boxes 23 1/16
₹19,700.00INR / Each
COMPARE
11 boxes 25 3/8
₹21,000.00INR / Each
COMPARE
12 boxes 27 11/16
₹22,800.00INR / Each
COMPARE
13 boxes 29 15/16
₹26,900.00INR / Each