Cole-Parmer® Vertical/Chest Aluminum Racks for Standard 3" Boxes
Aluminum racks offer better temperature uniformity in vapor phase in liquid nitrogen tanks
  • Lightweight aluminum construction for easier handling
  • Accommodates standard 3" cardboard, plastic, and metal boxes
  • Locking rod keeps sample storage boxes securely in the rack
Use these vertical racks with a wide variety of chest freezers and liquid nitrogen tanks. Dual handles make it easy to place racks in and out of freezers and nitrogen tanks.
16,000.00 - ₹26,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in)
Availability Pricing
COMPARE
6 boxes 19 1/16
₹16,000.00INR / Each
COMPARE
8 boxes 25 7/16
₹20,700.00INR / Each
COMPARE
9 boxes 28 1/4
₹24,000.00INR / Each
COMPARE
10 boxes 31 3/4
₹26,600.00INR / Each