Cole-Parmer® Variable Area Water Flowmeters, Brass Fittings, No Valves
Our variable area acrylic flowmeters offer precision and durability for a wide range of applications
  • Crystal-clear acrylic material with a stable float ensures easy viewing and long-lasting performance
  • Durable, corrosion-resistant brass fittings ensure robust connections for reliable use
  • Easily convertible from panel to partial or full in-line mounting configurations to suit your needs
  • Easy-to-read English or metric scales for clear and precise measurement
  • Panel mounting with hex nuts makes installation simple and secure
  • Easy disassembly and reassembly for quick cleaning and maintenance
Whether you need an accurate water flow measurement solution for industrial processes, lab setups, or HVAC systems, our acrylic flowmeters provide the perfect combination of precision, durability, and convenience. With a variety of customizable options, they are designed to fit seamlessly into your workflow. Flowmeters without valves are best suited for applications where the flow rate remains constant or is controlled by another device.
12,500.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Material Float Max Flow Rate (GPH) Max Flow Rate (mL/min)
Availability Pricing
COMPARE
Glass 0.3 20
₹12,500.00INR / Each
COMPARE
Stainless 1.1 70
₹12,500.00INR / Each
COMPARE
Carboloy 1.5 100
₹12,500.00INR / Each
COMPARE
Glass 2.75 175
₹12,500.00INR / Each
COMPARE
Stainless 6.5 450
₹12,500.00INR / Each
COMPARE
Carboloy 11 700
₹12,500.00INR / Each
COMPARE
Stainless 22 1400
₹12,500.00INR / Each
Showing 1- 7 of 7