Cole-Parmer® Variable-Area Panel-Mount Flowmeters, PTFE Fittings
Designed with unique rotatable scales for direct reading of air and water measurements
  • Graduations reflect both metric and English scales: L/min and scfm (air) and L/min and GPM (water)
  • Leak integrity rating of 1 x 10-7 sccs helium or better
  • PTFE fittings and float offer excellent chemical resistance
  • Panel mount for easy incorporation into an existing system
  • Compact, durable construction
  • ±5% of full-scale accuracy
  • Withstands temperatures up to 65°C (150°F)
Install these panel-mount flowmeters vertically inline to easily measure the flow rate of your system. Use the flowmeters with both air and water; rotatable flow tube features direct reading graduations in L/min and scfm for air measurements and L/min and GPM for water measurements. Each flowmeter is tested on a mass spectrometer leak detector and certified to a leak integrity rating of 1 x 10-7 sccs helium or better. Polycarbonate tube shield, heavy-walled borosilicate glass flow tube, PCTFE fittings, and PTFE float offer durability for long service life.
90,300.00 - ₹104,000.00INR / Each
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Max Air Flow Rate-L/min (LmPerin) Max Water Flow Rate-L/min (LmPerin) Valve
Availability Pricing
COMPARE
100 3 Yes
₹104,000.00INR / Each
COMPARE
200 5 3/4 Yes
₹104,000.00INR / Each
COMPARE
300 8 1/4 Yes
₹104,000.00INR / Each
COMPARE
400 11 Yes
₹104,000.00INR / Each
COMPARE
500 13 1/4 Yes
₹104,000.00INR / Each
COMPARE
625 16 Yes
₹104,000.00INR / Each
COMPARE
100 3 No
₹90,300.00INR / Each
COMPARE
200 5 3/4 No
₹90,300.00INR / Each
COMPARE
300 8 1/4 No
₹90,300.00INR / Each
COMPARE
400 11 No
₹90,300.00INR / Each
COMPARE
500 13 1/4 No
₹90,300.00INR / Each
COMPARE
625 16 No
₹90,300.00INR / Each
Showing 1- 12 of 12