Cole-Parmer® Variable-Area Panel-Mount Flowmeters, 316 SS Fittings
Designed with unique rotatable scales for direct reading of air and water measurements
  • Graduations reflect both metric and English scales: L/min and scfm (air) and L/min and GPM (water)
  • 316 SS fittings and float offer excellent chemical resistance
  • Panel mount for easy incorporation into an existing system
  • Compact, durable construction
  • ±5% of full-scale accuracy
  • Withstands temperatures up to 121°C (250°F)
Install these panel-mount flowmeters vertically inline to easily measure the flow rate of your system. Use the flowmeters with both air and water; rotatable flow tube features direct reading graduations in L/min and scfm for air measurements and L/min and GPM for water measurements. Polycarbonate tube shield, heavy-walled borosilicate glass flow tube, 316 SS fittings, and 316 SS float offer durability for long service life.
77,800.00 - ₹91,600.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Max Air Flow Rate-L/min (LmPerin) Max Water Flow Rate-L/min (LmPerin) Valve
Availability Pricing
COMPARE
140 4 Yes
₹91,600.00INR / Each
COMPARE
280 8 Yes
₹91,600.00INR / Each
COMPARE
425 11 1/2 Yes
₹91,600.00INR / Each
COMPARE
575 15 Yes
₹91,600.00INR / Each
COMPARE
900 20 Yes
₹91,600.00INR / Each
COMPARE
140 4 No
₹77,800.00INR / Each
COMPARE
280 8 No
₹77,800.00INR / Each
COMPARE
425 11 1/2 No
₹77,800.00INR / Each
COMPARE
575 15 No
₹77,800.00INR / Each
COMPARE
900 20 No
₹77,800.00INR / Each
Showing 1- 10 of 10