Cole-Parmer® Variable Area Flowmeters, 150 mm, Stainless Steel Mounting Fitting, High-Resolution Valve
Ideal for precise low flow rate measurements
  • Corrosion-resistant stainless-steel fittings ensure long-term reliability
  • Easy installation and exchange of flow tubes
  • Non-rotating adapter prevents tubes from turning
  • Panel mounting with hex nuts makes installation simple and secure
  • Chemical compatibility
  • 1 year warranty
These single flow tube variable area flowmeters with valves are designed for precise low flow rate measurements of air and water in mL/minute at a given set of pressure and temperature parameters. The Optigrad™ 150 mm universal millimeter scale minimizes parallax and eye fatigue. Particularly suitable for metering carrier gases in chromatography, indicating and controlling gases in manufacturing processes, liquid and gas measurement in laboratories, and pilot plants.
Built-in high-resolution control valves offer high sensitivity control and resolution in conjunction with metering tubes of very low flow rates. Other features include standard mounting fittings, side plates, thick protective magnifying front shield and back plate, and flow tubes.
91,700.00 - ₹93,600.00INR / Each
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Material Float Max Air Flow Rate (mLmin) Max Pressure (psig)
Availability Pricing
COMPARE
Sapphire 18 200
₹91,700.00INR / Each
COMPARE
Glass 46 200
₹91,700.00INR / Each
COMPARE
Glass 91 200
₹91,700.00INR / Each
COMPARE
Stainless 138 200
₹91,700.00INR / Each
COMPARE
Stainless 262 200
₹91,700.00INR / Each
COMPARE
Glass 370 200
₹91,700.00INR / Each
COMPARE
Stainless 816 200
₹91,700.00INR / Each
COMPARE
Stainless 1665 200
₹91,700.00INR / Each
COMPARE
Glass 2214 200
₹91,700.00INR / Each
COMPARE
Glass 3780 200
₹91,700.00INR / Each
COMPARE
Stainless 4494 200
₹91,700.00INR / Each
COMPARE
Glass 8555 200
₹91,700.00INR / Each
COMPARE
Stainless 16493 200
₹91,700.00INR / Each
COMPARE
Glass 23105 200
₹93,600.00INR / Each
COMPARE
Stainless 42860 200
₹93,600.00INR / Each
COMPARE
Carboloy 60212 200
₹93,600.00INR / Each
Showing 1- 16 of 16