Cole-Parmer® VapLock™ Adapter Fittings, Compression to Threaded NPT(M) with Large Bore for Semi-Rigid Tubing
Designed for secure leak-resistant connections
  • Constructed of durable polypropylene
Adapters with large bore for semi-rigid tubing are constructed of two-piece molded polypropylene components and do not include a separate ferrule. Adapters withstand pressures up to 100 psi. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
500.00 - ₹3,200.00INR / Pkg of 5
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Thread size NPT(M) (port 2) (in)
Availability Pricing
COMPARE
5/16 Polypropylene 1/4
₹1,600.00INR / Pkg of 5
COMPARE
3/8 Polypropylene 1/4
₹1,800.00INR / Pkg of 5
COMPARE
1/2 Polypropylene 1/4
₹3,200.00INR / Pkg of 5
COMPARE
3/8 Polypropylene 1/2
₹2,500.00INR / Pkg of 5
COMPARE
1/2 Polypropylene 1/2
₹2,700.00INR / Pkg of 5
COMPARE
1/2 Polypropylene 1/4
₹700.00INR / Each
COMPARE
3/8 Polypropylene 1/2
₹500.00INR / Each
Showing 1- 7 of 7