Cole-Parmer® Vacuum Desiccators with White Base, PP/PC
Holds vacuum up to 29” Hg for 24 hours
  • Knurled knob provides easy handling of dome
  • Autoclavable
These vacuum desiccators can hold a vacuum up to 29” (740 mm) Hg for 24 hours without any greasing. The top dome with molded-in knurled knob is constructed of rigid, transparent polycarbonate (PC), providing you a clear view of your sample.
The lower half is made of white polypropylene (PP) and includes a perforated PP desiccator plate. An internal groove on the flange holds the silicon rubber O-ring that provides leakproof vacuum capability.
A PP stopcock accepts ¼” (6.4 mm) ID vacuum tubing and features a self-lubricating PTFE plug. The three-way plug allows you to create, hold, and release the vacuum. Autoclavable at 121°C for 15 minutes.
22,800.00 - ₹38,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clearance Above Plate (mm) Flange Outside Diameter (OD) (cm) Material
Availability Pricing
COMPARE
123 17.1 Polycarbonate dome, Polypropylene base
₹27,000.00INR / Each
COMPARE
166 23.2 Polycarbonate dome, Polypropylene base
₹38,400.00INR / Each
COMPARE
207 27.6 Polycarbonate dome, Polypropylene base
₹22,800.00INR / Each
COMPARE
255 34 Polycarbonate dome, Polypropylene base
₹28,700.00INR / Each