Cole-Parmer® Upright Freezer Racks for 50-Cell Hinged Top Plastic Boxes
Get organized for easier retrieval of samples
  • Conveniently store 50-cell hinged top plastic boxes (2-1/4" x 3-3/8" x 5-9/16")
  • Included locking rods keep boxes securely in place
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors.
31,500.00 - ₹54,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
5 x 3 Array 7 18 3/8
₹31,500.00INR / Each
COMPARE
6 x 4 Array 9 5/16 22
₹37,500.00INR / Each
COMPARE
7 x 6 Array 13 15/16 25 11/16
₹54,000.00INR / Each