Cole-Parmer® Upright Freezer Drawer Racks for Large 3.75" High Boxes
Drawers slide out independently for easy access to the boxes
  • Use with large (3-3/4" x 5-13/16" x 5-13/16") boxes
  • Sturdy corrosion-resistant stainless steel construction for long-term use
Racks feature handles at both ends to accommodate freezers with right- or left-handed doors. Drawer racks facilitate access to samples without having to remove the entire rack from the freezer. Easily retrieve sample boxes by simply sliding out the individual rack drawer and then sliding the boxes out from either side of the rack.
26,000.00 - ₹46,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Height (in) Length (in)
Availability Pricing
COMPARE
3 x 2 Array 8 3/8 17 7/8
₹26,000.00INR / Each
COMPARE
4 x 3 Array 12 1/2 24
₹46,300.00INR / Each