Cole-Parmer® Essentials Universal Pipette Tips with Filter, Low Retention, Sterile
Protect against cross-contamination with tips free of human touch
  • Pure porous polyethylene filter protects against cross-contamination
  • Durable polypropylene construction
  • Noncytotoxic and nonhemolytic
  • Sterilized and DNase, RNase, and pyrogen free
Cole-Parmer sterile pipette tips are constructed of high-grade hydrophobic polypropylene and a polyethylene filter, which prevents liquids and aerosols from entering the body of your pipette. Low retention tips feature a hydrophobic and smooth surface. Tips are 99% Bacterial Aerosol Filtration Efficient (BFE), ASTM F21012, ISO 10993-5 MRM, minimum essential media elution, and GLP compliant.
23,600.00 - ₹27,400.00INR / Case of 960
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Filter Type Sterile Tip Volume (mL)
Availability Pricing
COMPARE
Polyethylene Yes 0.01
₹23,600.00INR / Case of 960
COMPARE
Polyethylene Yes 0.1
₹24,500.00INR / Case of 960
COMPARE
Polyethylene Yes 0.2
₹24,800.00INR / Case of 960
COMPARE
Polyethylene Yes 1
₹27,400.00INR / Case of 960
COMPARE
Polyethylene Yes 1.25
₹26,500.00INR / Case of 960
COMPARE
Polyethylene Yes 0.02
₹23,900.00INR / Case of 960