Cole-Parmer Ultraviolet Viewing Cabinets
Get uniform UV illumination for optimal viewing
  • Stay comfortable with soft rubber viewport viewing area
  • Removable bottom panel allows cabinet to be placed over large samples or transilluminator
  • Block out ambient light with lightweight access curtain
  • Protect your eyes with contrast control filter
These customizable UV viewing cabinets provide a compact darkroom environment for viewing materials under ultraviolet conditions. Manufactured of durable lightweight plastic, the cabinets are designed for long life and rugged use in a wide range of applications. Bottom panel of each cabinet is removable and the unit can easily be placed over samples or a transilluminator. A lightweight access curtain allows you easy entry and blocks out external light. The view port combines a comfortable soft rubber, which can accommodate eyeglasses, and a contrast control filter blocks out ultraviolet radiation.
A wide range of cabinet models and ultraviolet wavelength configurations for use in the laboratory or industrial applications are available.
61,600.00 - ₹569,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Power (watts) Width (in)
Availability Pricing
COMPARE
4 10 1/2
₹61,600.00INR / Each
COMPARE
230 8 19 3/4
₹408,000.00INR / Each
COMPARE
230 15 23 2/5
₹569,000.00INR / Each
COMPARE
230 15 23 2/5
₹527,000.00INR / Each
COMPARE
230 15 19 3/4
₹559,000.00INR / Each