Cole-Parmer® UC 200 Series Ultrasonic Cleaners with Digital Timer and Heat
High-frequency ultrasonic cleaning at an economical price
  • Multiple power settings handle both normal or delicate cleaning
  • Degas mode for improved cleaning efficiency and easy sample prep
  • Timed operation from 1-99 minutes
  • Adjustable heating to 176°F (80?) for deeper cleaning
Cole-Parmer ultrasonic cleaners are extremely effective at removing dirt and grime which would normally require tedious manual cleaning. The high frequency ultrasonic wave vibrations create microscopic bubbles that implode violently causing the cavitation which creates an intense scrubbing action on the surface of the item being cleaned. The microscopic nature of the bubbles mean even the tiniest crevices will be cleaned thoroughly. Used in a wide variety of industries and applications almost any non-porous item that can normally be immersed in water can be cleaned with these units.
These stainless steel cleaners feature a bright LED display for time and temperature and easy push-button control. Sturdy rubber feet prevent movement while in use, handles allow easy portability, and the larger sizes (6 liter and above) have external drains for easy replenishment of cleaning solutions.
45,600.00 - ₹181,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Gal) Power (VAC) Description
Availability Pricing
COMPARE
0.5 120 2 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹45,600.00INR / Each
COMPARE
0.5 230 2 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹45,800.00INR / Each
COMPARE
0.75 120 3 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹62,800.00INR / Each
COMPARE
0.75 230 3 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹62,200.00INR / Each
COMPARE
1.5 120 6 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹94,000.00INR / Each
COMPARE
1.5 230 6 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹93,000.00INR / Each
COMPARE
2.5 120 9 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹110,000.00INR / Each
COMPARE
2.5 230 9 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹109,000.00INR / Each
COMPARE
3.5 120 13 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹132,000.00INR / Each
COMPARE
3.5 230 13 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹131,000.00INR / Each
COMPARE
5.25 120 20 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹155,000.00INR / Each
COMPARE
5.25 230 20 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹153,000.00INR / Each
COMPARE
7.25 120 27 Liter Ultrasonic Cleaner with Digital Timer and Heat, 120 VAC
₹181,000.00INR / Each
COMPARE
7.25 230 27 Liter Ultrasonic Cleaner with Digital Timer and Heat, 230 VAC
₹177,000.00INR / Each
Showing 1- 14 of 14