Cole-Parmer® Ultra-Quiet Mini Lab Compressors
Compact, low-vibration, quiet compressed air stations
  • Get compressed air anywhere you need it, quietly—compact design and sound enclosure
  • Quick-connect port provides simple plug-and-play use of hoses and tools
  • Control outlet pressure with the turn of a dial-up to 70 psi (4.8 bar)
  • Manage maintenance easily-filters are easy to replace and have long service lives
These versatile lab compressors are designed for point-of-use applications—move station with the integrated handle or optional transport cart. The built-in sound enclosure allows for ultra-quiet operation, and the compact design and low-vibration performance allow placement anywhere needed. Quick-release coupling port comes with a 3/8" (9.5 mm) ID hose barb insert for easy tubing connection. Quickly see outlet pressure on the analog pressure gauge and use the control knob for adjustment up to 70 psi (4.8 bar)—ideal for cleaning dust off sensitive equipment and components. Maintenance is easy to manage—replace the air intake filter annually (bi-annually for high dust areas).
1,119,000.00 - ₹1,347,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Tank Size (Liters) Description
Availability Pricing
COMPARE
0.81 2 Cole-Parmer Ultra-Quiet 42 Mini Lab Compressor
₹1,119,000.00INR / Each
COMPARE
1.45 2 Cole-Parmer Ultra-Quiet 77 Mini Lab Compressor
₹1,347,000.00INR / Each