Cole-Parmer® Ultra Flex Support Systems with Bench Clamp
Ultra Flex support system offer the ultimate in flexibility for laboratory use
  • Flexible arms are extremely versatile and can be placed in virtually any position or angle
  • Designed to fit easily on benchtops or in fume hoods
Each Ultra Flex support system includes three interchangeable clamp heads: two-prong clamp head that opens to 2.6" ( 6.6 cm) dia, three-prong clamp head that opens to 2.3" (5.8 cm) dia, spring clamp head that opens to 0.5" (1.3 cm) dia and utility wrench.
Ultra Flex Support System with Bench Clamp
Bench clamp system includes flex arm, interchangeable clamp heads and bench/self clamp. . Designed to securely attach to the side of your lab bench or counter top, to help utilize more space in your lab. Bench clamp made of aluminum
96,200.00 - ₹114,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Clamp Type Length (in) Description
Availability Pricing
COMPARE
Equipment, Two- or Three-Prong 12 Ultra Flex Support System with 12" Flex Arm and Bench Clamp
₹96,200.00INR / Each
COMPARE
Equipment, Two- or Three-Prong 18 Ultra Flex Support System with 18" Flex Arm and Bench Clamp
₹114,000.00INR / Each