Cole-Parmer® Ultra Flex Support System with Laboratory Frame Connector
Ultra Flex support system offer the ultimate in flexibility for laboratory use
  • Flexible arms are extremely versatile and can be placed in virtually any position or angle
  • Designed to fit easily on benchtops or in fume hoods
Each Ultra Flex support system includes three interchangeable clamp heads: two-prong clamp head that opens to 2.6" ( 6.6 cm) dia, three-prong clamp head that opens to 2.3" (5.8 cm) dia, spring clamp head that opens to 0.5" (1.3 cm) dia and utility wrench.
Ultra Flex Support System with Lab-Frame Connector
Lab-frame system includes flex arm, interchangeable clamp heads and rod connector. Designed to securely attach to support stands, lab-frames, or any support rod up to 0.75” (19 mm) in diameter.
84,000.00 - ₹100,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Accepts Rod Diameter (in) Clamp Type Length (in)
Availability Pricing
COMPARE
0.75 Equipment, Two- or Three-Prong 12
₹84,000.00INR / Each
COMPARE
0.75 Equipment, Two- or Three-Prong 18
₹100,000.00INR / Each