Cole-Parmer® Ultra Flex Support System with Base Plate
Ultra Flex support system offer the ultimate in flexibility for laboratory use
  • Flexible arms are extremely versatile and can be placed in virtually any position or angle
  • Designed to fit easily on benchtops or in fume hoods
Each Ultra Flex support system includes three interchangeable clamp heads: two-prong clamp head that opens to 2.6" ( 6.6 cm) dia, three-prong clamp head that opens to 2.3" (5.8 cm) dia, spring clamp head that opens to 0.5" (1.3 cm) dia and utility wrench.
Ultra Flex Support System with base plate
Base plate system includes flex arm, interchangeable clamp heads and base plate. Designed to fit easily on bench tops or in fume hoods with a stable 5 x 5” (12.7 x 12.7 cm) chemical resistant black finish steel base plate.
108,000.00 - ₹130,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Clamp Type Length (in)
Availability Pricing
COMPARE
Includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head 12
₹108,000.00INR / Each
COMPARE
Includes a 2-Prong Clamp Head, 3-Prong Clamp Head, Spring Clamp Head 18
₹130,000.00INR / Each