Cole-Parmer® Essentials Ultra-Clean Premium Aluminum Foil
Eliminate problems caused by thinner foils—switch to premium foil
  • Impermeable to light, oxygen, and water
  • 100% recyclable
  • Handles extreme temperatures
  • Autoclavable
Cole-Parmer aluminum foil is manufactured specifically for the lab with added strength, puncture resistance, and purity. Foil is ideal for covering flasks, beakers, petri dishes, weigh boats, disposable crucibles, and for wrapping samples.
9,000.00 - ₹16,800.00INR / Case of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Width (in)
Availability Pricing
COMPARE
300 Standard aluminum 12
₹9,000.00INR / Case of 12
COMPARE
600 Heavy-duty aluminum 18
₹16,800.00INR / Case of 12
COMPARE
300 Extra-heavy-duty 18
₹14,200.00INR / Case of 12