Cole-Parmer® Two-Hole Stoppers, Rubber
These two-hole black stoppers made with solid natural rubber stay pliable over a long period of time. Provide good tightness and adhesion, withstand laboratory conditions without hardening, and are ideal for use with nonaggressive or low concentration solutions. Each stopper has its size marked in raised figures. Low sulfur content. Withstand temperatures from –13 to 158°F (–25 to 70°C).
8,600.00 - ₹17,300.00INR / Pkg of 63
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottom Outside Diameter (OD) (mm) Standard Size Number Description
Availability Pricing
COMPARE
10 00 Two-Hole Black Rubber Stoppers, Standard Size 00; 113/Pk
₹10,700.00INR / Pkg of 113
COMPARE
14 1 Two-Hole Black Rubber Stoppers, Standard Size 1; 63/Pk
₹10,900.00INR / Pkg of 63
COMPARE
18 3 Two-Hole Black Rubber Stoppers, Standard Size 3; 43/Pk
₹12,800.00INR / Pkg of 43
COMPARE
20 4 Two-Hole Black Rubber Stoppers, Standard Size 4; 37/Pk
₹12,300.00INR / Pkg of 37
COMPARE
23 5 Two-Hole Black Rubber Stoppers, Standard Size 5; 30/Pk
₹10,700.00INR / Pkg of 30
COMPARE
26 6 Two-Hole Black Rubber Stoppers, Standard Size 6; 22/Pk
₹10,000.00INR / Pkg of 22
COMPARE
27 6.5 Two-Hole Black Rubber Stoppers, Standard Size 6-1/2; 21/Pk
₹12,500.00INR / Pkg of 21
COMPARE
30 7 Two-Hole Black Rubber Stoppers, Standard Size 7; 16/Pk
₹8,600.00INR / Pkg of 16
COMPARE
33 8 Two-Hole Black Rubber Stoppers, Standard Size 8; 13/Pk
₹13,800.00INR / Pkg of 13
COMPARE
37 9 Two-Hole Black Rubber Stoppers, Standard Size 9; 11/Pk
₹13,400.00INR / Pkg of 11
COMPARE
42 10 Two-Hole Black Rubber Stoppers, Standard Size 10; 8/Pk
₹17,200.00INR / Pkg of 8
COMPARE
48 11 Two-Hole Black Rubber Stoppers, Standard Size 11; 6/Pk
₹17,300.00INR / Pkg of 6
Showing 1- 12 of 12