Cole-Parmer® Tuff-Tip, Gel-Filled, Side-Mount pH Electrodes
Durable Tuff-Tip with ability to mount sideways
  • No more breakage—tip design protects bulb
  • Full range; measure up to pH of 14
The tip design protects the bulb from breakage and the gel internal fill solution allows electrode to be installed where others cannot. Rugged PVDF housing with 1/2'' NPT (M) threads withstands temperatures from 23 to 176°F (--5 to 80°C) and pressures to 100 psi. Use for measurements up to pH 13. Models include a 10-ft cable with BNC pH connector. Insertion depth 1''.
40,700.00 - ₹46,200.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Side-Mount Tuff-Tip pH Probe, DJ/PVDF/No ATC; BNC
₹40,700.00INR / Each
COMPARE
Side-Mount Tuff-Tip pH Probe, DJ/PVDF/100Ohm RTD; Tinned/BNC
₹44,100.00INR / Each
COMPARE
Side-Mount Tuff-Tip pH Probe, DJ/PVDF/1kOhm RTD; Spade/BNC
₹45,800.00INR / Each
COMPARE
Side-Mount Tuff-Tip pH Probe, DJ/PVDF/100Ohm RTD; Tinned/Tinned
₹46,200.00INR / Each