Cole-Parmer® Tube Roller Racks
Expands your tube roller shaker capacity
  • Saves process time
  • Accommodates 10 tubes
  • Silicone rings offer smooth rotation
  • Tubes snap securely into holder for stable hold
  • Easily organize samples pre and post mixing
  • Autoclavable at 121 °C
Each tube roller rack can accommodate up to 10 tubes, doubling or tripling the capacity of your tube roller shaker. The rack saves you processing time by reducing the number of roller cycles. No need to invest in multiple roller shakers since the tube roller rack handles numerous tube samples at the same time.
The tube holder axis is parallel to roller axis for pure horizontal rotation for controlled sample manipulation. The racks allow you to slow the effective rpm of the tube for a gentler mix.
Use it as an upright storage tube rack or submerge it in an ice bath. Made of polypropylene material, the racks are wear-resistant and offer reliable sterilization through autoclaving.
23,100.00INR / Pkg of 3
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Tube Size (mL) Max Tube Size (mm)
Availability Pricing
COMPARE
11 15 20
₹23,100.00INR / Pkg of 3
COMPARE
9 2/5 7 17
₹23,100.00INR / Pkg of 3