Cole-Parmer® Transport Tubes, Sterile
Store, transport, or freeze samples in transport tubes
  • Made of high-grade polypropylene (PP)
  • White marking area makes it easy to label tubes
  • Easy-to-read white graduation markings
  • Sterile, RNase-free, DNase-free, and pyrogen-free
These flat-bottom, self-standing tubes make it easy to store and transport samples. High-grade PP construction offers durability even after being frozen. Each tube comes with a leak-proof HDPE screw-on cap. Packaged in 10 bags of 100 tubes.
38,700.00 - ₹44,600.00INR / Case of 1000
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tube Capacity (mL) Tube Length (mm)
Availability Pricing
COMPARE
5 60
₹38,700.00INR / Case of 1000
COMPARE
10 80
₹44,600.00INR / Case of 1000