Cole-Parmer® Threaded to Hose Barb Fitting, Straight Adapter, PP
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
  • Meets FDA requirements 21 CFR 177.1520(a)(3)(i) and (c)3.1a and RoHS compliant
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These animal-derivative-free polypropylene fittings are radiation stable and resistant to solvents, chemicals, water, strong mineral acids, bases, and other inorganic environments. Not recommended for use with oxidizing agents.
1,100.00 - ₹1,600.00INR / Pkg of 10
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Fitting Material Thread size UNF(M) (port 1) (InchShort)
Availability Pricing
COMPARE
1/8 Polypropylene 1/4"-28
₹1,500.00INR / Pkg of 10
COMPARE
5/32 Polypropylene 1/4"-28
₹1,600.00INR / Pkg of 10
COMPARE
1/16 Polypropylene 10-32
₹1,100.00INR / Pkg of 10
COMPARE
3/32 Polypropylene 10-32
₹1,100.00INR / Pkg of 10