Cole-Parmer® Threaded to Hose Barb Fitting, 90° Elbow, HDPE
Threaded barb adapter design provides safe, difficult-to-break connection
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Single barb design tightly “grips” the tubing
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other critical-use applications. Quickly and easily transition from soft tubing to a system that accepts a threaded connection using threaded barb adapters. Threads provide a tight and secure connection; a single barb on the other end of the fitting creates a tight seal to keep the tubing in place. An anti-rotation device on the hose barb prevents tubing from rotating, minimizing kinks. These high-density polyethylene (HDPE) fittings are softer than nylon, polypropylene, and Kynar®. They exhibit excellent resistance to chemicals but have lower thermal resistance than other materials.
1,800.00 - ₹2,700.00INR / Pkg of 10
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (in) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
COMPARE
1/8 1/8
₹1,800.00INR / Pkg of 10
COMPARE
5/32 1/8
₹1,800.00INR / Pkg of 10
COMPARE
3/16 1/8
₹1,800.00INR / Pkg of 10
COMPARE
1/4 1/8
₹1,800.00INR / Pkg of 10
COMPARE
3/8 1/8
₹1,800.00INR / Pkg of 10
COMPARE
1/8 1/4
₹1,800.00INR / Pkg of 10
COMPARE
5/32 1/4
₹1,800.00INR / Pkg of 10
COMPARE
3/16 1/4
₹1,800.00INR / Pkg of 10
COMPARE
1/4 1/4
₹1,800.00INR / Pkg of 10
COMPARE
5/16 1/4
₹1,800.00INR / Pkg of 10
COMPARE
3/8 1/4
₹1,800.00INR / Pkg of 10
COMPARE
1/2 1/4
₹1,800.00INR / Pkg of 10
COMPARE
1/4 3/8
₹1,800.00INR / Pkg of 10
COMPARE
5/16 3/8
₹1,900.00INR / Pkg of 10
COMPARE
3/8 3/8
₹1,900.00INR / Pkg of 10
COMPARE
1/2 3/8
₹1,900.00INR / Pkg of 10
COMPARE
1/4 1/2
₹1,900.00INR / Pkg of 10
COMPARE
3/8 1/2
₹1,900.00INR / Pkg of 10
COMPARE
1/2 1/2
₹1,900.00INR / Pkg of 10
COMPARE
3/4 1/2
₹2,200.00INR / Pkg of 10
Showing 1 - 20 of 28