Cole-Parmer® Threaded, In-Line Housings
Simply insert electrode—screws into tee fitting
  • Pipe tee connection is 1⁄2" NPT(M)
  • Viton O-ring seals electrode for waterproof protection
  • Housing measures 313⁄16" L x 1" dia (9.7 x 2.5 cm)
Simply slip an electrode into this housing and screw it into a tee fitting. Pipe tee connection is 1/2'' NPT(M). Includes a VITON O-ring. KYNAR construction provides a superior chemical resistance. Maximum pressure: 150 psi. Guarded models increase electrode life. KYNAR model is suitable for harsh chemicals. Stainless steel (SS) guard models is ideal for food and high-temperature applications. Housings measures 3-13/16''L x 1'' dia.
12,900.00 - ₹23,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
pH Electrode Housing, threaded/in-line, w/o bulb guard, PVDF
₹12,900.00INR / Each
COMPARE
Threaded In-Line Housing with Guard
₹23,100.00INR / Each