Cole-Parmer® Sugar Hydrometer, Brix, Plain form
  • NIST Traceable
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serializ
Used in the food and beverage and other industries for measuring % sucrose by weight, these plain form hydrometers are available in various ranges from -5° to 71° Bri
14,900.00 - ₹22,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/35 Degree Brix Sugar Scale Hydrometer
₹14,900.00INR / Each
COMPARE
0/12 Degree Brix Sugar Scale Hydrometer
₹15,300.00INR / Each
COMPARE
9/21 Degree Brix Sugar Scale Hydrometer
₹15,400.00INR / Each
COMPARE
19/31 Degree Brix Sugar Scale Hydrometer
₹22,300.00INR / Each