Cole-Parmer® Standard Single Quartz Cuvettes for UV Spectroscopy
Transmission over 80% at 200 nm
  • Standard-size cuvettes are the type most often used for routine laboratory testing
  • Cuvettes include chemical resistant, PTFE covers
  • Cuvettes have two clear sides
These Cole-Parmer® standard single quartz cuvettes are constructed of quartz and suitable for use in UV spectroscopy applications.
21,200.00 - ₹91,200.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Max Capacity (mL) Pathlength (mm)
Availability Pricing
COMPARE
Quartz 0.4 1
₹47,700.00INR / Each
COMPARE
Quartz 0.7 2
₹22,900.00INR / Each
COMPARE
Quartz 1.7 5
₹21,200.00INR / Each
COMPARE
Quartz 3.5 10
₹33,000.00INR / Each
COMPARE
Quartz 7 20
₹28,200.00INR / Each
COMPARE
Quartz 10.5 30
₹30,500.00INR / Each
COMPARE
Quartz 14 40
₹68,300.00INR / Each
COMPARE
Quartz 17.5 50
₹91,200.00INR / Each
Showing 1- 8 of 8