Cole-Parmer® Specific Gravity Plain Form Precision Hydrometers for Liquids Heavier than Water
  • Temperature of Standardization 60°F/60°F
  • Metal Ballast
  • Individually Serialized
Individually packaged with a multi-language Statement of Accuracy
9,200.00 - ₹12,700.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Precision 1.000/1.070 Specific Gravity Hydrometer
₹9,200.00INR / Each
COMPARE
Precision 1.060/1.130 Specific Gravity Hydrometer
₹11,200.00INR / Each
COMPARE
Precision 1.120/1.190 Specific Gravity Hydrometer
₹11,400.00INR / Each
COMPARE
Precision 1.180/1.250 Specific Gravity Hydrometer
₹11,400.00INR / Each
COMPARE
Precision 1.240/1.310 Specific Gravity Hydrometer
₹11,800.00INR / Each
COMPARE
Precision 1.300/1.370 Specific Gravity Hydrometer
₹12,200.00INR / Each
COMPARE
Precision 1.360/1.430 Specific Gravity Hydrometer
₹12,000.00INR / Each
COMPARE
Precision 1.420/1.490 Specific Gravity Hydrometer
₹12,600.00INR / Each
COMPARE
Precision 1.480/1.550 Specific Gravity Hydrometer
₹12,700.00INR / Each
Showing 1- 9 of 9