Cole-Parmer® SPE Micropipette Tips, All Carbon Reverse Phase
Porous carbon network—allows for efficient retention and separation of large molecules, highly polar, and closely related structures at extreme pHs
  • Improved removal of nuisance compounds and impurities
  • Reduce sample processing time with high throughput, low solvent usage
  • Promotes sustainability - can be reused, cleaned, and sanitized with alkaline washes
The Cole-Parmer® SPE all carbon reverse phase micropipette tips are suitable for sample preparation of small molecules, peptides, protein, RNA, DNA before chromatography, mass spectrometry, and PCR. Designed with a natural graphite all carbon source, these pipette tips facilitate optimal and reproducible extraction of analytes and impurities, improving quality control, regulatory compliance, and increasing chromatographic systems lifespan. This improved productivity and reduction of consumables can reduce operations and maintenance costs.
Choose from dispersive tips with loose media and two frits or monolith lips with a network of chained together microbeads and one frit.
41,900.00INR / Pkg of 96
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (µL) Bed Mass (mg) Methodology
Availability Pricing
COMPARE
1000 30 Dispersive micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
200 10 Dispersive micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
1000 20 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
200 5 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
10 1 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
1000 50 Dispersive micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
200 6 Dispersive micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
1000 40 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
200 2 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
COMPARE
10 0.5 Monolith fixed bed micro-SPE
₹41,900.00INR / Pkg of 96
Showing 1- 10 of 10