Cole-Parmer® SP Laboratory Overhead Mixer
Variable speed mixer designed for reliability and durability
  • Durable—no wear user serviceable parts Low/no maintanence brushless motor Adjustable tilt from 0 to 30 degrees allows proper impeller placement IP44 rated per IEC 60529 Hollow through shaft and user-replaceable keyless chuck
Cole-Parmer SP laboratory mixer ensures accurate, dependable motor speed control with “soft-start” feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimizes speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.
The mixer rotates at 50 to 2500 rpm with a maximum torque of 91 in-oz (64 N·cm) and has a through-shaft capability. Bracket hardware is preinstalled to enable installation of the adjustable tilt mounting bracket. The motor cable connector has a locking mechanism when the cable is connected to prevent unintended disconnection in the event of accidental force on the motor cable.
The variable-speed controller is used to turn on and set the speed and direction of the mixer motor. Easily switch the direction of rotation using the Forward-Off-Reverse switch. The LED indicates the mixer is powered by turning green and flashes when a limit condition has been reached.
454,000.00 - ₹571,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Min Speed (rpm) Description
Availability Pricing
COMPARE
2500 50 SP Mixer Set; 120/230 VAC
₹454,000.00INR / Each
COMPARE
2500 50 SP Mixer System; 120/230 VAC
₹571,000.00INR / Each