Cole-Parmer® Solution Bottles
Ideal for storing, transferring and media preparation
  • Unique, stable design to prevent accidental tip over
  • Light weight polystyrene solution bottles are available with 5 different volumes of 150, 250, 500, 1000, and 2000ml
  • Heavy wall construction and edge knurls on the cap for easy screw
  • Wide mouth bottles for stable and efficient pour out
  • Engraved or silk-printed graduation
  • Special concave for easy handling
  • Gamma sterilized
  • Single package in easy-open packs to assure free from contamination
  • Polypropylene bottle cap is individually packed
  • Inner packaging is individually labelled for lot traceability
6,400.00 - ₹21,900.00INR / Case of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Sterile Description
Availability Pricing
COMPARE
150 Yes 1 bottle/bag; 24 bag/case; Total: 24 bottles
₹6,400.00INR / Case of 1
COMPARE
250 Yes 1 bottle/bag; 24 bag/case; Total: 24 bottles
₹7,700.00INR / Case of 1
COMPARE
500 Yes 1 bottle/bag; 24 bag/case; Total: 24 bottles
₹8,500.00INR / Case of 1
COMPARE
1000 Yes 1 bottle/bag; 24 bag/case; Total: 24 bottles
₹21,900.00INR / Case of 1
COMPARE
2000 Yes 1 bottle/bag; 12 bag/case; Total: 12 bottles
₹11,100.00INR / Case of 1