Cole-Parmer® SHP-200 Series Large Capacity Infrared Stirring Hot Plates
Efficient infrared heater saves you time and energy
  • Large plate surface is ideal for heating large volumes of liquid
  • Ceramic top stays cool around the edges, preventing burns
  • Be safe—flashing light warns when top plate is hot
  • BioCote® antimicrobial protection is built in for a cleaner, more hygienic product
Efficient infrared heater combined with a stirrer delivers faster, evenly heated fluids. A 900-watt infrared heater can boil a liter of water over 30% faster than a conventional, 1200-watt ceramic hot plate. Rare earth magnets provide powerful stirring—up to 15 liters—and feedback control from 100 to 1500 rpm.
"Hot" warning light flashes when the plate temperature is above 50 °C and remains lit even when the unit is turned off.
Units feature BioCote antimicrobial protection which reduces the presence of odor and stain causing microbes such as bacteria, mold, and fungi on the product surface by up to 99.99%.
131,000.00 - ₹137,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
High Speed (rpm) Max Temperature (° F) Top Plate Length (in)
Availability Pricing
COMPARE
1500 842 12
₹131,000.00INR / Each
COMPARE
1500 842 12
₹137,000.00INR / Each