Cole-Parmer® Sanitary Pressure Transducers
The only "no fill" sanitary transducer available
  • ±0.1% full-scale accuracy
  • Designed for clean-in-place and steam-in-place installations
Sanitary pressure transducers are designed to meet 3A standards — fully sealed to withstand external high-pressure washdowns. Applications include food, dairy, pharmaceutical and paint manufacturing. As a totally self-contained electronic package, the capacitance sensing element along with the signal conditioned IC-based circuit ensures excellent accuracy and long-term stability.
407,000.00 - ₹425,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Process Connection Accuracy Max Pressure (PSI)
Availability Pricing
COMPARE
1-1/2" Tri-Clamp® ±0.1% full-scale 30
₹425,000.00INR / Each
COMPARE
1-1/2" Tri-Clamp® ±0.1% full-scale 60
₹407,000.00INR / Each
COMPARE
1-1/2" Tri-Clamp® ±0.1% full-scale 100
₹425,000.00INR / Each
COMPARE
2" Tri-Clamp® ±0.1% full-scale 150
₹407,000.00INR / Each