Cole-Parmer® SamplePrep Ring and Puck Mill Grinding Containers
Use with ShatterBox® Verastile Ring and Puck Mill to pulverizing and mix samples
  • Dish-shaped grinding container swings in a tight high-speed circle to crush samples to analytical fineness
Optimize performance of your Cole-Parmer® Shatterbox® Ring and Puck Mill with these grinding containers. Typical samples include cements, soils, ceramics, slag, rocks, minerals, ores, pharmaceuticals, sulfur pellets, and dried plant material.
121,000.00 - ₹1,601,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Material Description
Availability Pricing
Cole-Parmer SamplePrep 8501 Grinding Container, Hardened Steel, 270 mL; 1/EA 0457675 Mfr # 8501
COMPARE
270 Hardened steel Grinding Container, Hardened Steel, 270 mL; 1/EA
₹326,000.00INR / Each
Cole-Parmer SamplePrep 8504 Grinding Container, Tungsten Carbide Lined, 240 mL; 1/EA 0457676 Mfr # 8504
COMPARE
240 Tungsten carbide/steel Grinding Container, Tungsten Carbide Lined, 240 mL; 1/EA
₹1,601,000.00INR / Each
Cole-Parmer SamplePrep 8505 Grinding Container, Alumina Ceramic Lined, 170 mL; 1/EA 0457677 Mfr # 8505
COMPARE
170 Alumina ceramic/aluminum/steel Grinding Container, Alumina Ceramic Lined, 170 mL; 1/EA
₹482,000.00INR / Each
Cole-Parmer SamplePrep 8506 Grinding Container, Zirconia Lined, 170 mL; 1/EA 0457678 Mfr # 8506
COMPARE
170 Zirconia ceramic/SS/aluminum Grinding Container, Zirconia Lined, 170 mL; 1/EA
₹916,000.00INR / Each
Cole-Parmer SamplePrep 8507 Grinding Container, Hardened Steel, 90 mL; 1/EA 0457679 Mfr # 8507
COMPARE
90 Hardened steel Grinding Container, Hardened Steel, 90 mL; 1/EA
₹121,000.00INR / Each
Cole-Parmer SamplePrep 8508 Grinding Container, Tungsten Carbide, 80 mL; 1/EA 0457680 Mfr # 8508
COMPARE
80 Tungsten carbide Grinding Container, Tungsten Carbide, 80 mL; 1/EA
₹666,000.00INR / Each
Cole-Parmer SamplePrep 8521 Grinding Container, Hardened Steel, 650 mL; 1/EA 0457681 Mfr # 8521
COMPARE
650 Hardened steel Grinding Container, Hardened Steel, 650 mL; 1/EA
₹433,000.00INR / Each
Showing 1- 7 of 7