Cole-Parmer® Salt/Combination Analog Refractometers
Quick and easy operation with reliable measurements
  • Easy calibration via the calibration screw for enhanced accuracy
  • No more worrying about temperature fluctuations with the automatic temperature compensation (ATC) models
These units come in four range options, including salinity, brix, refractive index, and a combination of the these. For the units that feature automatic temperature compensation (ATC), these allow the user to concentrate on the measurements without worrying about the temperature. The compensation range is 50 to 86°F (10 to 30°C).
All models also feature a zero% adjust with distilled water. This allows for immediate and easy calibration for precise measurements at all times.
18,000.00 - ₹20,400.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max Salinity Range (%) Description
Availability Pricing
COMPARE
±0.5%; ±0.001 100 Refractometer 0 - 100%, Salinity
₹18,000.00INR / Each
COMPARE
±0.2% 28 Refractometer 0 - 28%, Salinity
₹20,400.00INR / Each
COMPARE
±0.5%; ±0.001; ±0.1% 100 Refractometer 0 - 100%, Salinity, 1.000 - 1.070 RI, 0 - 10% Brix
₹18,500.00INR / Each
COMPARE
±0.2% 28 Refractometer 0 - 28%, Salinity
₹18,500.00INR / Each
COMPARE
±0.5%; ±0.001 100 Refractometer w ATC, 0 - 100% Salinity, 1.000 - 1.070 RI
₹18,600.00INR / Each
COMPARE
±0.2% 28 Refractometer w ATC, 0 - 28%, Salinity
₹18,500.00INR / Each
COMPARE
±0.5%; ±0.001; ±0.1% 100 Refractometer w ATC, 0 - 100% Salinity, 1.000 - 1.070 RI, 0 - 10% Brix
₹18,500.00INR / Each
COMPARE
±0.2% 28 Refractometer w ATC, 0 - 28% Salinity, 0 - 32% Brix
₹18,500.00INR / Each
Showing 1- 8 of 8