Cole-Parmer® SAFETYBLUE Precision Brix Thermo-Hydrometers
  • NIST Traceable
  • Blue Spirit
  • Non-Mercury
  • Temperature of Standardization 20°C/20°C
  • Individually Serialized
These thermo-hydrometers feature non-mercury performance liquid and conform to the accuracy requirements for ASTM thermo-hydrometers. Each thermo-hydrometer is calibrated in a triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST. Made in the USA.
15,900.00 - ₹19,600.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
0/12 Degree Brix Sugar Scale Combined Form Thermo-Hydrometer
₹15,900.00INR / Each
COMPARE
9/21 Degree Brix Sugar Scale Combined Form Thermo-Hydrometer
₹15,900.00INR / Each
COMPARE
19/31 Degree Brix Sugar Scale Combined Form Thermo-Hydrometer
₹18,100.00INR / Each
COMPARE
29/41 Degree Brix Sugar Scale Combined Form Thermo-Hydrometer
₹16,900.00INR / Each
COMPARE
59/71 Degree Brix Sugar Scale Combined Form Thermo-Hydrometer
₹19,600.00INR / Each