Cole-Parmer® S-Connectors
Connect rods and / or clamps to your frame or build a custom lab frame
  • Use with 1/2" (12.7mm) diameter frame rods
This connector allows for the connecting of two perpendicular rods, ideal for use when constructing lab-frames or clamp supports. Connects two ¨1/2" (13mm) rods at a 90° angle and features separate adjustment screws for each rod location.
12,500.00 - ₹17,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accepts Rod Diameter (in) Clamp Material Clamp Type
Availability Pricing
COMPARE
0.5 Stainless Steel Frame and Rod
₹12,500.00INR / Each
COMPARE
0.5 Nickel-plated Zinc Frame and Rod
₹17,000.00INR / Each