Cole-Parmer® RV-200 Series Rotary Evaporators
Simple one-handed lift operation for ease of use
  • Set and view rotation speed, temperature, and timer on two large LCDs
  • A unique PTFE sealing system offers excellent thermostability
  • PID temperature controller provides stable temperature control
  • Small footprint takes up minimal lab space
Easily set and view operating parameters on the two large digital displays; one for rotation speed and timer and another for bath temperature and timer. A unique PTFE sealing system provides great thermostability and excellent chemical resistance. A stainless steel bath with PTFE coating withstands high temperatures and is easy to clean. Safety features include dry-run protection, overtemperature shutoff, circuit breaker, and power interruption alarm.
268,000.00 - ₹508,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Evaporation Flask Size Lift Mechanism Power (VAC)
Availability Pricing
COMPARE
2 L* Manual 120
₹391,000.00INR / Each
COMPARE
2 L* Manual 220
₹268,000.00INR / Each
COMPARE
2 L* Motorized 120
₹508,000.00INR / Each
COMPARE
2 L* Motorized 220
₹402,000.00INR / Each