Cole-Parmer® PVC 3-Stop Pump Tubing
Cost effective PVC tubing offers excellent durability and reliable flow rates
  • Stops ensure a perfect fit every time
  • Chemically resistant to a wide range of solutions
  • Delivers accurate and interference-free analytical results
  • Color-coded stops for easy identification
  • Transparent—easily view fluid flow
  • Certified to USP Class VI, USP37-NF32 2014
  • Meets European Pharmacopoeia Monograph 3.1.2
  • Ideal for general-purpose, laboratory, and single-use applications
  • Free technical application support available to assist with tubing selection
Made from phthalate-free PVC, the tubing exhibits great resistance to a wide range of common chemicals, ensuring dependable performance for many routine applications. Anti-leaching characteristics minimize the risk of interferences or false positives, ensuring accurate and reliable results. Tubing is transparent so you can easily observe the fluid flow. PVC tubing withstands temperatures ranging from -50 to 74°C (-58 to 165°F).
5,000.00 - ₹6,000.00INR / Pkg of 12
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Coded Stops Tubing ID (mm)
Availability Pricing
COMPARE
Orange/Red 0.19
₹6,000.00INR / Pkg of 12
COMPARE
Orange/Blue 0.25
₹5,900.00INR / Pkg of 12
COMPARE
Orange/Green 0.38
₹5,000.00INR / Pkg of 12
COMPARE
Green/Yellow 0.44
₹5,000.00INR / Pkg of 12
COMPARE
Orange/Yellow 0.51
₹5,000.00INR / Pkg of 12
COMPARE
Orange/White 0.64
₹5,000.00INR / Pkg of 12
COMPARE
Black/Black 0.76
₹5,000.00INR / Pkg of 12
COMPARE
Orange/Orange 0.89
₹6,000.00INR / Pkg of 12
COMPARE
White/White 1.02
₹5,000.00INR / Pkg of 12
COMPARE
Red/Red 1.14
₹6,000.00INR / Pkg of 12
COMPARE
Gray/Gray 1.3
₹5,000.00INR / Pkg of 12
COMPARE
Yellow/Yellow 1.42
₹6,000.00INR / Pkg of 12
COMPARE
Yellow/Blue 1.52
₹5,000.00INR / Pkg of 12
COMPARE
Blue/Blue 1.65
₹6,000.00INR / Pkg of 12
COMPARE
Green/Green 1.85
₹6,000.00INR / Pkg of 12
COMPARE
Purple/Purple 2.06
₹6,000.00INR / Pkg of 12
COMPARE
Purple/Orange 2.54
₹5,000.00INR / Pkg of 12
COMPARE
Purple/White 2.79
₹5,000.00INR / Pkg of 12
Showing 1- 18 of 18