Cole-Parmer® PTFE Vials
PTFE material offers almost total chemical resistance
  • Non-stick, leading to easy recovery of contents
  • Stable over a wide temperature range
Cole-Parmer PTFE vials feature almost total chemical resistance to almost any chemical compound in a laboratory environment. Additionally, PTFE material is stable over a wide temperature range from -200°C to 300°C making it suitable for a wide range of applications. Material is high purity and totally inert preventing contamination. Vial material is non-stick for easy sample transfer and recovery.
20,900.00 - ₹27,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Volume (mL)
Availability Pricing
COMPARE
25
₹20,900.00INR / Each
COMPARE
50
₹27,300.00INR / Each