Cole-Parmer® PTFE Syringe Filters
Unaffected by most organic solvents, concentrated acids and bases, propellants, and cryogenic fluids
  • Ideal for sterilization of gas and alcohols; cleaning acids, alkalis, and organic solvents; venting purposes.
  • Use with aggressive chemicals
Polytetrafluoroethylene (PTFE) membranes are Triton-free, have a bidirectional membrane support, one threaded end and one barbed end (50 mm dia) for a secure fit to the syringe. Membrane type and pore size are printed on each filter for easy identification. The syringe filters are 100% integrity tested and manufactured in accordance with ISO 9002 standards. They are certified nonpyrogenic, noncytotoxic, and are blister packed.
6,500.00INR / Pkg of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Membrane Diameter (mm) Pore Size (µm) Sterile
Availability Pricing
COMPARE
50 0.45 No
₹6,500.00INR / Pkg of 10