Cole-Parmer® PTFE Needle Valves
Provides precise regulation of corrosive gasses and liquids
  • Fluids contact PTFE and PCTFE only
  • Structurally Rigid Metal Shell
  • One PTFE o-ring
  • Simplicity only seven components
27,900.00 - ₹63,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Max Flow Rate For Air (mL/min) Max Flow Rate For Water (mL/min)
Availability Pricing
COMPARE
1/8" NPT(F) 2400 130
₹27,900.00INR / Each
COMPARE
1/8" NPT(F) 55000 2800
₹27,900.00INR / Each
COMPARE
1/4" Compression 2400 130
₹63,400.00INR / Each
COMPARE
1/4" Compression 55000 2800
₹63,400.00INR / Each