Cole-Parmer® PTFE Metering Valves
Provides precise regulation of corrosive gasses and liquids
  • Non-fluid contacting external parts are made of anodized aluminum
  • Valves offer three conveniently overlapping flow ranges
  • Safety handle prevents over tightening and facilitates fine metered regulation
23,500.00 - ₹42,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connection Type Max Flow Rate For Air (mL/min) Max Flow Rate For Water (mL/min)
Availability Pricing
COMPARE
1/8" NPT(F) 600 36
₹23,500.00INR / Each
COMPARE
1/8" NPT(F) 3000 180
₹23,500.00INR / Each
COMPARE
1/8" NPT(F) 30000 1800
₹23,500.00INR / Each
COMPARE
1/4" Compression 600 36
₹42,000.00INR / Each
COMPARE
1/4" Compression 3000 180
₹42,000.00INR / Each
COMPARE
1/4" Compression 30000 1800
₹42,000.00INR / Each