Cole-Parmer® PTFE Dipper
PTFE material offers almost total chemical resistance
  • Nonstick surface leads to easy recovery of contents
  • Stable over a wide temperature range
  • Material is FDA Compliant and USP Class VI approved
Cole-Parmer PTFE dippers feature almost total chemical resistance to most chemical compounds. Additionally, PTFE material is stable from -328 to 572°F (-200°C to 300°C), making it suitable for a wide range of applications. Material is high purity and totally inert preventing contamination. Shaft is PTFE-encapsulated steel; extra shaft can extend length. Dipper can be sterilized via autoclave or gamma irradiation.
33,200.00 - ₹52,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Overall Length (in)
Availability Pricing
COMPARE
100 24
₹33,200.00INR / Each
COMPARE
250 24
₹40,300.00INR / Each
COMPARE
500 24
₹46,400.00INR / Each
COMPARE
1 24
₹52,700.00INR / Each