Cole-Parmer® PTFE-Coated Fiberglass Tapes
Tapes are made from woven fiberglass fabric and have silicone adhesive on one side. Tapes have excellent chemical, heat, and moisture resistance. Use for packaging, heat sealing, or shrink wrapping. Approved by USDA and is FDA compliant for food contact. Withstand temperatures from –100 to 500°F (–73 to 260 C).
5,400.00 - ₹37,200.00INR / Roll of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Thickness (mil) Width (in)
Availability Pricing
COMPARE
54 5 1/2
₹5,400.00INR / Roll of 1
COMPARE
54 5 1
₹9,600.00INR / Roll of 1
COMPARE
54 5 2
₹18,700.00INR / Roll of 1
COMPARE
54 8 1
₹14,000.00INR / Roll of 1
COMPARE
54 8 2
₹26,600.00INR / Roll of 1
COMPARE
54 12 2
₹37,200.00INR / Roll of 1