Cole-Parmer® PTFE Adhesive
Excellent electrical and thermal operating characteristics
  • Temperature range is -99 to 500°F (-73 to 260°C)
PTFE film (5 mil thick) features silicone adhesive backing. Use this tape to provide a nonstick, chemically inert surface or wrap.
13,800.00 - ₹84,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Thickness (mil) Width (in)
Availability Pricing
COMPARE
108 5 1/2
₹13,800.00INR / Each
COMPARE
108 5 1
₹24,000.00INR / Each
COMPARE
108 5 2
₹84,800.00INR / Each