Cole-Parmer® Powder Spatulas, Sterile, PS
Push the open-tipped, "U"-style blade into the powder to take a sample
  • Manufactured and packed in Class 100,000 cleanroom to ensure cleanliness and reduce cross contamination
  • Rounded handle makes it easy to hold
  • Made from high impact polystyrene (HIPS) which conforms to FDA, EC, and EU regulations
  • BSE/TSE free
The single-use powder spatulas are gamma irradiated and individually packaged so they can be used straight out of the bag without cleaning.
32,300.00 - ₹45,900.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Handle Length (InchShort) Overall Length (cm) Sterile
Availability Pricing
COMPARE
3 1/8 23 Yes
₹32,300.00INR / Pkg of 100
COMPARE
3 1/8 33 Yes
₹45,900.00INR / Pkg of 100